Learn More
Cayman Chemical AB-CHMINACA metabolite M6, Purity: ≥98%, CAS: 2748161-80-8, 1 mg
SDP
Catalog No. 502970412
Encompass_Preferred
A potential metabolite of AB-CHMINACA in which the isopropyl group has been oxidized; intended for research and forensic applications
Catalog No. 50-297-0412 Supplier Cayman Chemical Supplier No. 164531 MG
A potential metabolite of AB-CHMINACA in which the isopropyl group has been oxidized; intended for research and forensic applications