Learn More
Enzo Life Sciences IMMUNOSET alphaB-Crystallin ELISA development set (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI960074
Encompass_Preferred
The IMMUNOSET αB-Crystallin EIA development set contains sufficient reagents for the development of 5 x 96 well EIA plates which provides analysis of ~200 samples in duplicate. Save time, money, and sample by developing your own EIA kits. Simply coat your plates and go! This kit provides superior accuracy with fully quantitative results compared to Western blot analysis and has low reactivity with related molecules. Alternative name: CRYAB, HSPB5. Assay Time: Plate Coating - Overnight + 1 hour; Assay - 3 hours. Applications: ELISA, Colorimetric detection .
Catalog No. 50-200-7126