Learn More
Cayman Chemical AB-FUBINACA metabolite 2A, Purity: ≥98%, CAS: 2460433-24-1, 5 mg
SDP
Catalog No. 502969511
Encompass_Preferred
An expected carboxy metabolite of AB-FUBINACA, based on the major metabolites produced from similar synthetic CBs in humans; intended for forensic and research applications
Catalog No. 50-296-9511 Supplier Cayman Chemical Supplier No. 155295 MG
An expected carboxy metabolite of AB-FUBINACA, based on the major metabolites produced from similar synthetic CBs in humans; intended for forensic and research applications