Learn More
Amsbio LLC Ab-Heparan Sulfate (JM-403 epitope) / Rat, (JM-403) (mo) (M), Purified
Catalog No. 502566533
Encompass
Heparan sulfate antibody (clone JM403) reacts with an epitope present in many types of heparan sulfate. The JM403 epitope includes (an) N-unsubstituted glucosamine residue(s) that are critical for the reactivity of the antibody. The reactivity of the JM403 antibody with most heparan sulfates is nearly completely abolished after treatment of the glycosaminoglycan with bacterial Heparitinase I (Heparinase III) from Flavobacterium heparinum (EC 4.2.2.8). The JM403 antibody does not react with heparin, hyaluronan, chondroitin sulfate, dermatan sulfate and keratan sulfate.
Catalog No. 50-256-6533 Supplier Amsbio LLC Supplier No. 3707301
Heparan sulfate antibody (clone JM403) reacts with an epitope present in many types of heparan sulfate. The JM403 epitope includes (an) N-unsubstituted glucosamine residue(s) that are critical for the reactivity of the antibody. The reactivity of the JM403 antibody with most heparan sulfates is nearly completely abolished after treatment of the glycosaminoglycan with bacterial Heparitinase I (Heparinase III) from Flavobacterium heparinum (EC 4.2.2.8). The JM403 antibody does not react with heparin, hyaluronan, chondroitin sulfate, dermatan sulfate and keratan sulfate.