Learn More
Eurofins Discoverx AssayComplete Protein Dilution Buffer, 2 x 50. (92-0023M)
Catalog No. NC0900507
Encompass
AssayComplete Protein Dilution Buffer is a specially formulated dilution buffer designed for use in reconstituting protein agonists and diluting other ligands (antibodies, small molecules, etc.) to the proper stock concentration for use in Eurofins DiscoverX cell-based assays. Contains 10 plates and lids. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.
Catalog No. NC0900507 Supplier Eurofins Discoverx Supplier No. 920023M
AssayComplete Protein Dilution Buffer is a specially formulated dilution buffer designed for use in reconstituting protein agonists and diluting other ligands (antibodies, small molecules, etc.) to the proper stock concentration for use in Eurofins DiscoverX cell-based assays. Contains 10 plates and lids. For detailed product information, User manual and datasheet, visit www.discoverx.com/reagents.