Learn More
STEMCELL Technologies Anti-Mouse CD8a Antibody, Clone 53-6.7, FITC, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 60023FI.1
Encompass
Rat monoclonal IgG2a antibody against mouse, toad CD8a, FITC-conjugated
Catalog No. NC1662800