Learn More
Innovative Research Inc Pooled Human AB Serum Off The Clot Heat Inactivated 100ml
Catalog No. 502036398
Encompass_Preferred
Pooled Normal Human AB Serum Off The Clot and Heat Inactivated from Innovative Research is collected as whole blood donations from male donors with blood type AB. The blood is allowed to spontaneously clot and the serum component is extracted. The units are then pooled, filtered, bottled, and cell culture tested for mycoplasma, endotoxin, and sterility. The serum then undergoes a heat inactivation process to destroy complement activity. This product is suitable for 
Catalog No. 50-203-6398 Supplier Innovative Research Inc Supplier No. ISERABOTCHI100ML
Pooled Normal Human AB Serum Off The Clot and Heat Inactivated from Innovative Research is collected as whole blood donations from male donors with blood type AB. The blood is allowed to spontaneously clot and the serum component is extracted. The units are then pooled, filtered, bottled, and cell culture tested for mycoplasma, endotoxin, and sterility. The serum then undergoes a heat inactivation process to destroy complement activity. This product is suitable for 