Learn More
Takara Bio AB0481TAR HS DNA POL 250 UNIT
SDP
Catalog No. 50444006
Encompass
Enzyme; Polymerase; DNA; Speedstar HS; 250U; Used for high speed amplification and robust amplification of large DNA templates
Catalog No. 50-444-006 Supplier Takara Bio Supplier No. RR070A
Enzyme; Polymerase; DNA; Speedstar HS; 250U; Used for high speed amplification and robust amplification of large DNA templates