Learn More
Medchemexpress LLC HY-17423 10mg Medchemexpress, Abacavir CAS:136470-78-5 Purity:>98%
SDP
Catalog No. 502081948
Encompass_Preferred
Medchemexpress, HY-17423 10mg Abacavir CAS:136470-78-5 Abacavir is a potent nucleoside analog reverse-transcriptase inhibitor (NRTI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1948 Supplier Medchemexpress LLC Supplier No. HY1742310MG
Medchemexpress, HY-17423 10mg Abacavir CAS:136470-78-5 Abacavir is a potent nucleoside analog reverse-transcriptase inhibitor (NRTI). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.