Learn More
Medchemexpress LLC Abacavir | 136470-78-5 | 286.34 | 5 MG
SDP
Catalog No. 5000383993
Encompass_Preferred
Abacavir is an orally active and competitive nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and demonstrates anticancer activity in prostate cancer cell lines. Abacavir is also capable of trespassing the blood-brain-barrier and suppressing telomerase activity.
  • Orally active and competitive nucleoside reverse transcriptase inhibitor
  • Inhibits the replication of HIV
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity
Catalog No. 50-003-83993 Supplier Medchemexpress LLC Supplier No. HY174235MG
Abacavir is an orally active and competitive nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and demonstrates anticancer activity in prostate cancer cell lines. Abacavir is also capable of trespassing the blood-brain-barrier and suppressing telomerase activity.
  • Orally active and competitive nucleoside reverse transcriptase inhibitor
  • Inhibits the replication of HIV
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity