Learn More
Medchemexpress LLC Abacavir | 136470-78-5 | 99.0% | 286.34 | 10 MM 1 ML
SDP
Catalog No. 5000384604
Encompass_Preferred
Abacavir is an orally active and competitive nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and shows anticancer activity in prostate cancer cell lines. Abacavir is also capable of trespassing the blood-brain-barrier and suppressing telomerase activity.
  • Orally active
  • Competitive nucleoside reverse transcriptase inhibitor
  • Inhibits HIV replication
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity
Catalog No. 50-003-84604 Supplier Medchemexpress LLC Supplier No. HY1742310MM/1ML
Abacavir is an orally active and competitive nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and shows anticancer activity in prostate cancer cell lines. Abacavir is also capable of trespassing the blood-brain-barrier and suppressing telomerase activity.
  • Orally active
  • Competitive nucleoside reverse transcriptase inhibitor
  • Inhibits HIV replication
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity