Learn More
Selleck Chemical LLC Abacavir 25mg 136470-78-5
SDP
Supplier:  Selleck Chemical LLC S521525MG
Encompass_Preferred
Abacavir (ABC) is a powerful nucleoside analog reverse transcriptase inhibitor (NRTI) used to treat HIV and AIDS. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8325