Learn More
Medchemexpress LLC Abacavir hydroxyacet 100mg | 1446418-48-9 | 100MG
SDP
Catalog No. 5000240077
Encompass_Preferred
Abacavir hydroxyacetate is an orally active inhibitor of IL-20 and PRINS Abacavir hydroxyacetate reduces proliferation rate of skin in psoriasis as an immune modulator Abacavir hydroxyacetate can be used for the research of inflammatory skin problems[1 [2
Catalog No. 50-002-40077 Supplier Medchemexpress LLC Supplier No. HY106647100MG
Abacavir hydroxyacetate is an orally active inhibitor of IL-20 and PRINS Abacavir hydroxyacetate reduces proliferation rate of skin in psoriasis as an immune modulator Abacavir hydroxyacetate can be used for the research of inflammatory skin problems[1 [2