Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abacavir hydroxyacetate | 1446418-48-9 | 99.6% | 344.37 | C16H20N6O3 | 50 MG
SDP

Catalog No. 5000240222
Encompass_Preferred

Abacavir hydroxyacetate is a research-grade small molecule described as an orally active inhibitor of IL-20 and PRINS. It is used in studies of inflammatory skin conditions, including psoriasis, and is supplied as a high-purity solid in milligram quantities for laboratory research.

  • Orally active inhibitor of IL-20 and PRINS.
  • Used in research on inflammatory skin conditions, including psoriasis.
  • Molecular formula C16H20N6O3.
  • Molecular weight 344.37.
  • CAS RN 1446418-48-9.
  • Purity 99.6%.
  • Available in small-mass vials (e.g., 50 MG).

Catalog No. 50-002-40222 Supplier Medchemexpress LLC Supplier No. HY10664750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Abacavir hydroxyacetate is a research-grade small molecule described as an orally active inhibitor of IL-20 and PRINS. It is used in studies of inflammatory skin conditions, including psoriasis, and is supplied as a high-purity solid in milligram quantities for laboratory research.

  • Orally active inhibitor of IL-20 and PRINS.
  • Used in research on inflammatory skin conditions, including psoriasis.
  • Molecular formula C16H20N6O3.
  • Molecular weight 344.37.
  • CAS RN 1446418-48-9.
  • Purity 99.6%.
  • Available in small-mass vials (e.g., 50 MG).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.