Learn More
Medchemexpress LLC Abacavir hydroxyacetate | 1446418-48-9 | MFCD34470006 | 99.6% | 344.37 | C16H20N6O3 | 5 MG
SDP
Catalog No. 5000240224
Encompass_Preferred
Abacavir hydroxyacetate is a research-grade small molecule reported to be an orally active inhibitor of IL-20 and PRINS. It modulates immune responses and reduces skin cell proliferation in psoriasis models, making it suitable for studies of inflammatory skin disorders and pathway validation.
  • Orally active inhibitor of IL-20 and PRINS.
  • Useful for researching inflammatory skin disorders and psoriasis biology.
  • Provides a characterized small-molecule tool for pathway and target validation.
  • High stated purity (~99.6%) for reproducible experimental results.
  • Available in small research pack sizes for preclinical studies.
Catalog No. 50-002-40224 Supplier Medchemexpress LLC Supplier No. HY1066475MG
Abacavir hydroxyacetate is a research-grade small molecule reported to be an orally active inhibitor of IL-20 and PRINS. It modulates immune responses and reduces skin cell proliferation in psoriasis models, making it suitable for studies of inflammatory skin disorders and pathway validation.
  • Orally active inhibitor of IL-20 and PRINS.
  • Useful for researching inflammatory skin disorders and psoriasis biology.
  • Provides a characterized small-molecule tool for pathway and target validation.
  • High stated purity (~99.6%) for reproducible experimental results.
  • Available in small research pack sizes for preclinical studies.