Learn More
TARGETMOL CHEMICALS INC Abacavir sulfate 100MG
SDP
Catalog No. 502880147
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Abacavir sulfate (Ziagen) , a nucleoside analogue, effectively utilize its antiviral activity Purity 99.85%
Catalog No. 50-288-0147 Supplier TARGETMOL CHEMICALS INC Supplier No. T6367100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Abacavir sulfate (Ziagen) , a nucleoside analogue, effectively utilize its antiviral activity Purity 99.85%