Learn More
Medchemexpress LLC Abacavir sulfate | 188062-50-2 | 100.0% | 335.38 | 50 MG
SDP
Catalog No. 5000385645
Encompass_Preferred
Abacavir sulfate (Abacavir Hemisulfate) is a competitive, orally active nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and shows anticancer activity in prostate cancer cell lines. It is also able to trespass the blood-brain-barrier and suppress telomerase activity.
  • Inhibits replication of HIV
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity
Catalog No. 50-003-85645 Supplier Medchemexpress LLC Supplier No. HY17423A50MG
Abacavir sulfate (Abacavir Hemisulfate) is a competitive, orally active nucleoside reverse transcriptase inhibitor. It can inhibit the replication of HIV and shows anticancer activity in prostate cancer cell lines. It is also able to trespass the blood-brain-barrier and suppress telomerase activity.
  • Inhibits replication of HIV
  • Shows anticancer activity in prostate cancer cell lines
  • Can trespass the blood-brain-barrier
  • Suppresses telomerase activity