Learn More
Medchemexpress LLC Abacavir sulfate | 188062-50-2 | 99.9% | 335.38 | 5 MG
SDP
Catalog No. 5000386049
Encompass_Preferred
Abacavir sulfate (Abacavir Hemisulfate) is a competitive, orally active nucleoside reverse transcriptase inhibitor. It inhibits the replication of HIV, shows anticancer activity in prostate cancer cell lines, can trespass the blood-brain-barrier, and suppresses telomerase activity.
  • Competitive, orally active nucleoside reverse transcriptase inhibitor.
  • Inhibits the replication of HIV.
  • Shows anticancer activity in prostate cancer cell lines, inhibiting cell growth, affecting cell cycle progression, inducing senescence, and modulating LINE-1 mRNA expression.
  • Can trespass the blood-brain-barrier.
  • Suppresses telomerase activity.
  • Significantly reduces cell migration and inhibits cell invasion.
  • Induces fat apoptosis.
  • Enhances survival of high-risk medulloblastoma-bearing mice (when administered with Decitabine).
Catalog No. 50-003-86049 Supplier Medchemexpress LLC Supplier No. HY17423A5MG
Abacavir sulfate (Abacavir Hemisulfate) is a competitive, orally active nucleoside reverse transcriptase inhibitor. It inhibits the replication of HIV, shows anticancer activity in prostate cancer cell lines, can trespass the blood-brain-barrier, and suppresses telomerase activity.
  • Competitive, orally active nucleoside reverse transcriptase inhibitor.
  • Inhibits the replication of HIV.
  • Shows anticancer activity in prostate cancer cell lines, inhibiting cell growth, affecting cell cycle progression, inducing senescence, and modulating LINE-1 mRNA expression.
  • Can trespass the blood-brain-barrier.
  • Suppresses telomerase activity.
  • Significantly reduces cell migration and inhibits cell invasion.
  • Induces fat apoptosis.
  • Enhances survival of high-risk medulloblastoma-bearing mice (when administered with Decitabine).