Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abacavir sulfate, 99 %

Catalog No. AC458860010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC458860010 1 g
AC458860050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC458860010 Supplier Thermo Scientific Chemicals Supplier No. 458860010
Add to Cart

CAS: 188062-50-2 | C14H18N61/2H2SO4 | 335.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188062-50-2
Molecular Formula C14H18N61/2H2SO4
Molecular Weight (g/mol) 335.37
InChI Key WMHSRBZIJNQHKT-NHMRBCHTSA-N
Synonym abacavir hemisulfate, 1s,4r-4-2-amino-6-cyclopropylamino-9h-purin-9-yl-2-cyclopentene-1-methanol hemisulfate
PubChem CID 75539558
IUPAC Name [(1S)-4-[2-amino-6-(cyclopropylamino)purin-9-yl]cyclopent-2-en-1-yl]methanol;sulfuric acid
SMILES C1CC1NC2=NC(=NC3=C2N=CN3C4CC(C=C4)CO)N.C1CC1NC2=NC(=NC3=C2N=CN3C4CC(C=C4)CO)N.OS(=O)(=O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 335.37
Percent Purity 99%
Chemical Name or Material Abacavir sulfate
Hazard Category Serious eye damage/eye irritation (category 1), Germ cell mutagenicity (category 2), Reproductive toxicity (category 2), Carcinogenicity (category 2), Skin sensitisation (category 1), Hazardous to the aquatic environment (chronic category 3)
Hazard Statement Causes serious eye damage, Suspected of causing genetic defects, Suspected of damaging the unborn child, Suspected of causing cancer, May cause an allergic skin reaction, Harmful to aquatic life with long lasting effects

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.