Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abacavir 98.0+%, TCI America™
SDP

Catalog No. A26941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26941G 1 g
A26945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26941G Supplier TCI America Supplier No. A26941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 136470-78-5
Molecular Formula C14H18N6O
Molecular Weight (g/mol) 286.339
MDL Number MFCD00903850
InChI Key MCGSCOLBFJQGHM-SCZZXKLOSA-N
Synonym (1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-methanol
PubChem CID 441300
ChEBI CHEBI:421707
IUPAC Name [(1S,4R)-4-[2-amino-6-(cyclopropylamino)purin-9-yl]cyclopent-2-en-1-yl]methanol
SMILES C1CC1NC2=NC(=NC3=C2N=CN3C4CC(C=C4)CO)N

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 1 g
Formula Weight 286.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Abacavir
RTECSNumber GY5979200
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.