Learn More
Cayman Chemical Abacavr-d4 1mg
SDP
Catalog No. 502989968
Encompass_Preferred
An internal standard for the quantification of abacavir by GC- or LC-MS
Catalog No. 50-298-9968 Supplier Cayman Chemical Supplier No. 300761 MG
An internal standard for the quantification of abacavir by GC- or LC-MS