Learn More
Medchemexpress LLC HY-108742A 5mg Medchemexpress, Abaloparatide (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY108742A5MG
Encompass_Preferred
Medchemexpress, HY-108742A 5mg Abaloparatide (TFA) CAS: Abaloparatide TFA (BA 058 TFA) is a parathyroid hormone receptor 1 (PTHR1) analogue selected to be a potent and selective activator of the PTHR1 signaling pathway. Abaloparatide TFA enhances Gs/cAMP signaling (EC50 of 0.3 nM) and β-arrestin recruitment (EC50 of 0.9 nM) in MC3T3-E1 osteoblast cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8626