Learn More
Crescent Chemical Co Inc ABAMECTIN SOLN 1 ML
SDP
Supplier:  Crescent Chemical Co Inc PSXA10001000AL
Encompass_Preferred
Abamectine 100 µg/mL in Acetonitrile Insecticide (Metabolite neat) CAS 71751-41-2 Molecular Weight - 1732.1272 Storage - RT Catalog Num - DRE-XA10001000AL pesticide
Catalog No. 50-798-07