Learn More
Medchemexpress LLC Abametapir | 1762-34-1 | 184.24 | 25 G
SDP
Catalog No. 5000449982
Encompass_Preferred
Abametapir is a metalloproteinase (MMP) inhibitor that targets metalloproteinases crucial for egg hatching and louse development. It can inhibit the hatching of both head and body lice. Abametapir is also capable of chelating heavy metal ions such as iron, copper, and zinc, allowing it to interact with various insect targets that require metal co-factors for their function, including metalloproteinases.
  • Inhibits metalloproteinases critical for egg hatching and louse development.
  • Effective against both head and body lice.
  • Chelates heavy metal ions like iron, copper, and zinc.
  • Interacts with insect targets requiring metal co-factors.
Catalog No. 50-004-49982 Supplier Medchemexpress LLC Supplier No. 25GHYW004546
Abametapir is a metalloproteinase (MMP) inhibitor that targets metalloproteinases crucial for egg hatching and louse development. It can inhibit the hatching of both head and body lice. Abametapir is also capable of chelating heavy metal ions such as iron, copper, and zinc, allowing it to interact with various insect targets that require metal co-factors for their function, including metalloproteinases.
  • Inhibits metalloproteinases critical for egg hatching and louse development.
  • Effective against both head and body lice.
  • Chelates heavy metal ions like iron, copper, and zinc.
  • Interacts with insect targets requiring metal co-factors.