Learn More
TARGETMOL CHEMICALS INC ABAPERIDONE 5MG
SDP
Catalog No. 502581310
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Abaperidone is a potent 5-HT2A receptor and dopamine D2 receptor antagonist with an IC50 of 6.2 for 5-HT2A receptors and 17 nM for dopamine D2 receptors.Abaperidone is a potentially atypical anti-psychotropic compound that reduces basal hsp70 mRNA expression in rat striatum and prefrontal cortex. expression in the striatum and prefrontal cortex of rats. purity: 94%
Catalog No. 50-258-1310 Supplier TARGETMOL CHEMICALS INC Supplier No. T140835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Abaperidone is a potent 5-HT2A receptor and dopamine D2 receptor antagonist with an IC50 of 6.2 for 5-HT2A receptors and 17 nM for dopamine D2 receptors.Abaperidone is a potentially atypical anti-psychotropic compound that reduces basal hsp70 mRNA expression in rat striatum and prefrontal cortex. expression in the striatum and prefrontal cortex of rats. purity: 94%