Learn More
Aviva Systems Biology Corp ABAT Antibody - middle region (ARP51765_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51765_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ABAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ABAT. Alternate names: GABA-AT, GABAT, NPD009ApplicationWB, IHC. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60608