Learn More
Medchemexpress LLC ABAT Human His SU 50ug
SDP
Catalog No. 5000196302
Encompass_Preferred
Recombinant human ABAT (γ-aminobutyrate aminotransferase) produced in E. coli and supplied as a lyophilized powder. The protein includes N-terminal SUMO and 10xHis tags plus a C-terminal Myc tag, is approximately 69 kDa, and is >90% pure by reducing SDS-PAGE. Formulated from PBS with 6% trehalose (pH 7.4), it is intended for biochemical and cellular research on GABA metabolism and mitochondrial function; store frozen and follow reconstitution stability guidelines.
  • Expressed in E. coli with SUMO, 10xHis, and Myc tags for purification and detection.
  • High purity (>90%) suitable for biochemical assays.
  • Lyophilized formulation with trehalose for improved stability.
  • Approximate molecular weight 69 kDa for experimental planning.
  • Stable short term at 4°C after reconstitution; long-term storage at -20°C/-80°C when aliquoted.
Catalog No. 50-001-96302 Supplier Medchemexpress LLC Supplier No. HYP7206850UG
Recombinant human ABAT (γ-aminobutyrate aminotransferase) produced in E. coli and supplied as a lyophilized powder. The protein includes N-terminal SUMO and 10xHis tags plus a C-terminal Myc tag, is approximately 69 kDa, and is >90% pure by reducing SDS-PAGE. Formulated from PBS with 6% trehalose (pH 7.4), it is intended for biochemical and cellular research on GABA metabolism and mitochondrial function; store frozen and follow reconstitution stability guidelines.
  • Expressed in E. coli with SUMO, 10xHis, and Myc tags for purification and detection.
  • High purity (>90%) suitable for biochemical assays.
  • Lyophilized formulation with trehalose for improved stability.
  • Approximate molecular weight 69 kDa for experimental planning.
  • Stable short term at 4°C after reconstitution; long-term storage at -20°C/-80°C when aliquoted.