Learn More
Medchemexpress LLC Abatacept | 332348-12-6 | 99.01% | 84.4 kDa | 10 MG
SDP
Catalog No. 5000344694
Encompass_Preferred
Abatacept (powder) is a soluble fusion protein consisting of the extra-cellular domain of human CTLA4 and a fragment of the Fc portion of human IgG1 (hinge and CH2 and 3 domains). It is a selective T-cell co-stimulation modulator and a protein agent for autoimmune diseases.
  • Reduces paw edema.
  • Exhibits linear pharmacokinetics (PK) across studied doses.
  • Reduces the proportion of activated T cells (CD44highCD62L-) and inhibits the up-regulation of ICOS and CD71 in homozygous DO11.10 RAG-2-/- BALB/c (H-2d/d) mice.
Catalog No. 50-003-44694 Supplier Medchemexpress LLC Supplier No. HY108829A1MG
Abatacept (powder) is a soluble fusion protein consisting of the extra-cellular domain of human CTLA4 and a fragment of the Fc portion of human IgG1 (hinge and CH2 and 3 domains). It is a selective T-cell co-stimulation modulator and a protein agent for autoimmune diseases.
  • Reduces paw edema.
  • Exhibits linear pharmacokinetics (PK) across studied doses.
  • Reduces the proportion of activated T cells (CD44highCD62L-) and inhibits the up-regulation of ICOS and CD71 in homozygous DO11.10 RAG-2-/- BALB/c (H-2d/d) mice.