Learn More
Medchemexpress LLC Abaucin hydrochloride | 1173022-16-6 | >98% | 426.86 | 1 ML
SDP
Catalog No. 5000387795
Encompass_Preferred
Abaucin hydrochloride is a potent CCR2 antagonist (IC50: 360 nM) with no effect on CCR1, primarily used in neurological disease and endocrinology research. It targets human α1a, α1d receptors, and rat brain cortex 5HT1a receptor, and suppresses wild type and D284N mutant MCP-1 receptor activity. In vivo studies show it decreases pain thresholds in rats with bone cancer pain.
  • Potent CCR2 antagonist (IC50: 360 nM), no effect on CCR1
  • Inhibits α1a, α1d, and 5HT1a receptors
  • Suppresses wild type and D284N mutant MCP-1 receptor
  • Reduces extracellular matrix protein expression
  • Blocks fibronectin and type IV collagen expression in high glucose-stimulated mesangial cells
  • Abrogates increased TGF-1 levels in mesangial cells
  • Decreases pain threshold in rats with bone cancer pain
  • Reverses NR2B, nNOS, and SIGIRR expression in spinal cord
  • Used in neurological disease and endocrinology research
Catalog No. 50-003-87795 Supplier Medchemexpress LLC Supplier No. HY1861110MM/1ML
Abaucin hydrochloride is a potent CCR2 antagonist (IC50: 360 nM) with no effect on CCR1, primarily used in neurological disease and endocrinology research. It targets human α1a, α1d receptors, and rat brain cortex 5HT1a receptor, and suppresses wild type and D284N mutant MCP-1 receptor activity. In vivo studies show it decreases pain thresholds in rats with bone cancer pain.
  • Potent CCR2 antagonist (IC50: 360 nM), no effect on CCR1
  • Inhibits α1a, α1d, and 5HT1a receptors
  • Suppresses wild type and D284N mutant MCP-1 receptor
  • Reduces extracellular matrix protein expression
  • Blocks fibronectin and type IV collagen expression in high glucose-stimulated mesangial cells
  • Abrogates increased TGF-1 levels in mesangial cells
  • Decreases pain threshold in rats with bone cancer pain
  • Reverses NR2B, nNOS, and SIGIRR expression in spinal cord
  • Used in neurological disease and endocrinology research