Learn More
Anton Paar Abbemat Advanced refractometers deliver precise, consistent results for busy labs. Follow SOPs, integrate into multi-parameter systems, and use autosamplers to reduce errors. Seamless IT integration enables fast, efficient workflows.
Catalog No. 5000580497
Encompass_Preferred
Accuracy: ±0.0001 nD / ± 0.05 °BrixMeasuring range: 1.33 nD to 1.55 nD / 0 °Brix to 100 °BrixPrecise temperature control: 20 °C to 40 °COver 200 pre-installed measuring scalesIT network integration, automation, multi-parameter systems
Catalog No. 50-005-80497 Supplier Anton Paar Supplier No. 313365
Accuracy: ±0.0001 nD / ± 0.05 °BrixMeasuring range: 1.33 nD to 1.55 nD / 0 °Brix to 100 °BrixPrecise temperature control: 20 °C to 40 °COver 200 pre-installed measuring scalesIT network integration, automation, multi-parameter systems