Learn More
Anton Paar Abbemat Advanced refractometers deliver precise, consistent results for busy labs. Follow SOPs, integrate into multi-parameter systems, and use autosamplers to reduce errors. Seamless IT integration enables fast, efficient workflows.
Supplier:  Anton Paar 313367
Encompass_Preferred
Accuracy: ±0.00002 nD / ± 0.015 °BrixMeasuring range: 1.26 nD to 1.72 nD / 0 °Brix to 100 °BrixPrecise temperature control: 4 °C to 105 °COver 200 pre-installed measuring scalesIT network integration, automation, multi-parameter systems
Catalog No. 50-005-80498
Request a Quote
Please contact your sales representative for a quote or further ordering instructions.