Learn More
Anton Paar Abbemat Advanced refractometers deliver precise, consistent results for busy labs. Follow SOPs, integrate into multi-parameter systems, and use autosamplers to reduce errors. Seamless IT integration enables fast, efficient workflows.
Catalog No. 5000580550
Encompass_Preferred
Accuracy: ±0.00004 nD / ± 0.03 °BrixMeasuring range: 1.30 nD to 1.66 nD / 0 °Brix to 100 °BrixPrecise temperature control: 10 °C to 70 °COver 200 pre-installed measuring scalesIT network integration, automation, multi-parameter systems
Catalog No. 50-005-80550 Supplier Anton Paar Supplier No. 313366
Accuracy: ±0.00004 nD / ± 0.03 °BrixMeasuring range: 1.30 nD to 1.66 nD / 0 °Brix to 100 °BrixPrecise temperature control: 10 °C to 70 °COver 200 pre-installed measuring scalesIT network integration, automation, multi-parameter systems