Learn More
Anton Paar Abbemat Advanced refractometers deliver precise, consistent results for busy labs. Follow SOPs, integrate into multi-parameter systems, and use autosamplers to reduce errors. Seamless IT integration ensures data integrity and efficient workflow
Catalog No. 5000580459
Encompass_Preferred
Accuracy (nD): ±0.0001 nD / ± 0.05 °BrixMeasuring range (nD): 1.33 nD to 1.55 nD / 0 °Brix to 100 °BrixPrecise temperature control: 20 °C to 40 °CAudit-proof compliance and data integrity solutionsComprehensive qualification documentation package
Catalog No. 50-005-80459 Supplier Anton Paar Supplier No. 313368
Accuracy (nD): ±0.0001 nD / ± 0.05 °BrixMeasuring range (nD): 1.33 nD to 1.55 nD / 0 °Brix to 100 °BrixPrecise temperature control: 20 °C to 40 °CAudit-proof compliance and data integrity solutionsComprehensive qualification documentation package