Learn More
Abbott Piccolo Basic Metabolic Panel
Catalog No. 07P0204A Shop All Abbott Laboratories Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
07-P02-04A 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 07-P02-04A Supplier Abbott Supplier No. 07P0204
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Type Basic Metabolic Panel
For Use With (Application) For in vitro quantitative determination of: BUN, Ca2+, CRE, GLU, Na+, K+, tCO2, Cl-
Quantity 10/Pk.