Learn More
Abbott Piccolo Comprehensive Metabolic Panel
Catalog No. 07P0208A Shop All Abbott Laboratories Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
07-P02-08A 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 07-P02-08A Supplier Abbott Supplier No. 07P0208
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifications
Type Comprehensive Metabolic Panel
For Use With (Application) For in vitro quantitative determination of: ALB, ALP, ALT, AST, TBIL, TP, BUN, Ca2+, Cl-, CRE, GLU, K+, Na+, tCO2
Quantity 10/Pk.