Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ABBV-167 | 1351456-78-4 | 98.1% | C46H53ClN7O11PS | 1 ML
SDP

Catalog No. 5000354379
Encompass_Preferred

ABBV-167 is a phosphate prodrug of the BCL-2 inhibitor venetoclax. It demonstrated a lower food effect compared to venetoclax tablets and extensively converts to venetoclax following oral administration. This product is for research use only and has not been fully validated for medical applications.

  • Phosphate prodrug of BCL-2 inhibitor venetoclax
  • Demonstrates lower food effect compared to venetoclax tablets
  • Extensively converts to venetoclax after oral administration
  • Molecular weight: 978.45
  • Targets Bcl-2 family
  • Involved in Apoptosis pathway
  • Soluble in DMSO at 100 mg/mL (102.20 mM) with ultrasonication
  • In vivo solubility of ≥ 2.5 mg/mL (2.56 mM) in 10% DMSO and 90% (20% SBE-β-CD in saline)

Catalog No. 50-003-54379 Supplier Medchemexpress LLC Supplier No. HY14220910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABBV-167 is a phosphate prodrug of the BCL-2 inhibitor venetoclax. It demonstrated a lower food effect compared to venetoclax tablets and extensively converts to venetoclax following oral administration. This product is for research use only and has not been fully validated for medical applications.

  • Phosphate prodrug of BCL-2 inhibitor venetoclax
  • Demonstrates lower food effect compared to venetoclax tablets
  • Extensively converts to venetoclax after oral administration
  • Molecular weight: 978.45
  • Targets Bcl-2 family
  • Involved in Apoptosis pathway
  • Soluble in DMSO at 100 mg/mL (102.20 mM) with ultrasonication
  • In vivo solubility of ≥ 2.5 mg/mL (2.56 mM) in 10% DMSO and 90% (20% SBE-β-CD in saline)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.