Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ABBV-318 | 1802848-94-7 | 98.7% | 405.35 | 1 ML
SDP

Catalog No. 5000359819
Encompass_Preferred

ABBV-318 is a potent Nav1.7/Nav1.8 blocker, primarily used in research related to pain. It is characterized by high purity and specific activity against human Nav1.7 and Nav1.8 channels.

  • Potent Nav1.7/Nav1.8 blocker
  • IC50 for hNav1.7: 2.8 μM
  • IC50 for hNav1.8: 3.3 μM
  • Molecular weight: 405.35
  • Formula: C20H15F4N3O2
  • Appearance: White to light yellow solid
  • Purity: 98.7%

Catalog No. 50-003-59819 Supplier Medchemexpress LLC Supplier No. HY14606910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABBV-318 is a potent Nav1.7/Nav1.8 blocker, primarily used in research related to pain. It is characterized by high purity and specific activity against human Nav1.7 and Nav1.8 channels.

  • Potent Nav1.7/Nav1.8 blocker
  • IC50 for hNav1.7: 2.8 μM
  • IC50 for hNav1.8: 3.3 μM
  • Molecular weight: 405.35
  • Formula: C20H15F4N3O2
  • Appearance: White to light yellow solid
  • Purity: 98.7%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.