Learn More
TARGETMOL CHEMICALS INC ABBV-318 5MG
SDP
Catalog No. 502579491
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABBV-318 can be used as small molecule Nav1.7/Nav1.8 blockers for the treatment of pain with IC50 values of 2.8 uM for hNav1.7 and 3.8 uM for hNav1.8. ABBV-318 can be used to study pain-related disorders. purity: 99%
Catalog No. 50-257-9491 Supplier TARGETMOL CHEMICALS INC Supplier No. T620005MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABBV-318 can be used as small molecule Nav1.7/Nav1.8 blockers for the treatment of pain with IC50 values of 2.8 uM for hNav1.7 and 3.8 uM for hNav1.8. ABBV-318 can be used to study pain-related disorders. purity: 99%