Learn More
TARGETMOL CHEMICALS INC ABBV-744 10MG
SDP
Catalog No. 502579852
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABBV-744 is a BDII-selective BET bromodomain inhibitor that inhibits BRD2/3/4. It is used in the research on inflammatory diseases cancer and AIDS. purity: 99%
Catalog No. 50-257-9852 Supplier TARGETMOL CHEMICALS INC Supplier No. T469710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ABBV-744 is a BDII-selective BET bromodomain inhibitor that inhibits BRD2/3/4. It is used in the research on inflammatory diseases cancer and AIDS. purity: 99%