Learn More
Medchemexpress LLC ABBV-744 | 2138861-99-9 | 100.0% | 491.55 | 100 MG
SDP
Catalog No. 5000437723
Encompass_Preferred
ABBV-744 is a first-in-class, orally active, and selective inhibitor of the BDII domain of BET family proteins. It has IC50 values ranging from 4 to 18 nM for BRD2, BRD3, BRD4, and BRDT. It is primarily metabolized by CYP3A4, and its agent-like properties enable the investigation of its antitumor efficacy and tolerability. It is for research use only.
  • First-in-class
  • Orally active
  • Selective inhibitor of the BDII domain of BET family proteins
  • Exhibits antitumor efficacy and tolerability
Catalog No. 50-004-37723 Supplier Medchemexpress LLC Supplier No. HY112090100MG
ABBV-744 is a first-in-class, orally active, and selective inhibitor of the BDII domain of BET family proteins. It has IC50 values ranging from 4 to 18 nM for BRD2, BRD3, BRD4, and BRDT. It is primarily metabolized by CYP3A4, and its agent-like properties enable the investigation of its antitumor efficacy and tolerability. It is for research use only.
  • First-in-class
  • Orally active
  • Selective inhibitor of the BDII domain of BET family proteins
  • Exhibits antitumor efficacy and tolerability