Learn More
Selleck Chemical LLC ABBV-744 2mg 2138861-99-9
SDP
Supplier:  Selleck Chemical LLC S87232MG
Encompass_Preferred
ABBV-744 is a BDII-selective BET bromodomain inhibitor that inhibits BRD2, BRD3 and BRD4. It is developed for treating AML and cancers. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8275