Learn More
Medchemexpress LLC ABBV-CLS-484 1 MG
SDP
Supplier:  Medchemexpress LLC HY1459231MG
Encompass_Preferred
Osunprotafib (ABBV-CLS-484) is a potent PTPN1 and PTPN2 inhibitor with subnanomolar activity. Osunprotafib has antitumor activity, enhances the immune response and increases the sensitivity of tumors to immune-mediated killing.
Catalog No. NC2592110