Saint-Gobain 1 L single-use bioprocess bag designed for buffer and media storage, bioreactor feed and harvest, and intermediate product hold. The 2D bag features three ports with 20 in (50.8 cm) tubing and integrated fittings for sampling, transfer, and flow control. Multilayer film construction and Clear C-Flex 374 tubing provide broad chemical compatibility and a wide operating temperature range for downstream, upstream, and storage workflows.
- Volume: 1 L.
- Pack quantity: 20 per box.
- Ports: 3 ports with 20 in (50.8 cm) tubing on all ports.
- Port fittings: two ports with MPC insert, port cover, and pinch clamp; one port with LL female needle-free sampling port, luer cap, and pinch clamp.
- Overall dimensions: 13.50 x 6.48 in (34.29 x 16.46 cm).
- Film thickness: 7 mil for 50 mL-1 L bags.
- Tubing material: Clear C-Flex 374.
- Temperature range: -80 to +60 °C.