Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amerex™ ABC Dry Chemical Fire Extinguishers

Catalog No. 19041563 Shop All Amerex Corporation Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Discharge Time:
30 sec.
Material:
Aluminum
Model:
A411
UL Rating:
10A:120B:C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Discharge Time Material Model UL Rating
19-041-563 30 sec. Aluminum A411 10A:120B:C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-041-563 Supplier Amerex™ Supplier No. 11213
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rugged design with all metal valve construction

ABC extinguishers utilize a specially fluidized and siliconized mono ammonium phosphate dry chemical which chemically insulates Class A fires by melting at approximately 350°F and coats surface to which it is applied. It smothers and breaks the chain reaction of Class B fires and will not conduct electricity back to the operator.

  • Stored pressure design
  • Dependable drawn steel cylinders
  • Durable high gloss polyester powder paint
  • Anodized aluminum valve body
  • Anodized aluminum handle and lever
  • Temperature range: -65°F to 120°F

Specifications

Product Type Fire Extinguisher
Material Aluminum
Dimensions (L x W x H) 7 x 10.3 x 23.3 in. (17.7 x 26 x 59.1 cm)
Discharge Time 30 sec.
Height (English) 23.3 in.
Height (Metric) 59.1 cm
Includes Wall bracket
Length (English) 7 in.
Length (Metric) 17.7 cm
Model A411
Range 15 to 21 ft.
Reagent Type ABC dry chemical
Temperature Range (English) -65° to +120°F
Temperature Range (Metric) -53.8°C to +48.8°C
UL Rating 10A:120B:C
Weight (English) 20 lb.
Weight (Metric) 9.07 kg
Width (English) 10.3 in.
Width (Metric) 26 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.