Learn More
Vector Laboratories VECTASTAIN ABC-HRP (Peroxidase, Anti-Goat IgG) Kit
SDP
Catalog No. NC9664939
Encompass
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. The VECTASTAIN ABC kit system is modular and different variations can accommodate a wide array of primary antibody and tissue species. This kit uses horseradish peroxidase (HRP) as a detection enzyme and includes a secondary antibody that targets goat IgG primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation. Includes: 3 ml normal rabbit serum, 1 ml biotinylated rabbit anti-goat IgG secondary antibody, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated HRP). Stains 1000-2000 sections.
Catalog No. NC9664939 Supplier Vector Laboratories Supplier No. PK4005
With more than 50,000 citations, Vector Laboratories ABC kits are trusted by immunohistochemistry (IHC) and immunocytochemistry (ICC) laboratories. The VECTASTAIN ABC kit system is modular and different variations can accommodate a wide array of primary antibody and tissue species. This kit uses horseradish peroxidase (HRP) as a detection enzyme and includes a secondary antibody that targets goat IgG primary antibodies. The ABC complex, once formed, is stable and can be used for several days after preparation. Includes: 3 ml normal rabbit serum, 1 ml biotinylated rabbit anti-goat IgG secondary antibody, 2 ml Reagent A (avidin) and 2 ml Reagent B (biotinylated HRP). Stains 1000-2000 sections.