Learn More
Electron Microscopy Sciences ABC Lupe
SDP
Catalog No. 501902630
Encompass_Preferred
The Peak ABC Lupe is a precision folding lupe, consisting of A lens (12X), B lens (3X) and C lens (6X). With these three lenses combined, seven kinds of magnifying power are available. The lens is made from high quality glass optics and the body is made of aluminum. The magnification desired will be available from the combination table shown on the back of the lupe.
Catalog No. 50-190-2630 Supplier Electron Microscopy Sciences Supplier No. 68160
The Peak ABC Lupe is a precision folding lupe, consisting of A lens (12X), B lens (3X) and C lens (6X). With these three lenses combined, seven kinds of magnifying power are available. The lens is made from high quality glass optics and the body is made of aluminum. The magnification desired will be available from the combination table shown on the back of the lupe.