Learn More
ABC Vial™ LVI™, Sold by MilliporeSigma™ Supelco™
Catalog No. 111009864 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
Glass
Glass, PTFE
Glass, Silicone
Color:
Amber
Clear
Includes:
Cap, Vial
Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Color Includes
11-100-9864 Glass Clear Vial
11-100-9752 Glass, PTFE Amber Vial
11-100-9629 Glass, Silicone Clear Cap, Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-9864 Supplier MilliporeSigma Supelco Supplier No. 27562U
Add to Cart
Add to Cart
These ABC Vials contain a Low Volume Insert; can be used either as crimp seal or with screw cap.
Autosampler compatibility:
  • Agilent
  • Agilent 1050/1090A
  • Agilent 1100/1200
  • Agilent 7673A, 7683
  • Agilent 5890/7985A/6850/6890
  • LEAP Technologies
  • CTC LC PAL, GC PAL
  • CombiPAL
  • HTC PAL/HTS PAL
  • Shimadzu LC Autosamplers
  • SIL-HTa/SIL-HTc
  • SIL-10ADvp
  • SIL-10A/SIL-10Ai/SIL-10AP
  • Varian
  • CP-9010
  • 8000/8035/8100/8200
  • 9100/9090/9095
  • Vista
  • Waters
  • Alliance 2690/2695
  • Alliance 2790/2795
  • Acquity
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Glass
Includes Vial
No. per Pack 100
Volume (English) 0.3 mL
ABC Vial and LVI are trademarks of Wheaton Industries, Inc.