Learn More
Selleck Chemical LLC Opaganib (ABC294640) 5mg 915385-81-8
SDP
Supplier:  Selleck Chemical LLC S71745MG
Encompass_Preferred
Opaganib (ABC294640) is an orally bioavailable and selective sphingosine kinase-2 (SphK2) inhibitor with IC50 of approximately 60 ?M. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4485