Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Abc99 | 2331255-53-7 | 98.5% | 1 MG
SDP

Catalog No. 5000231459
Encompass_Preferred

ABC99 is an N-hydroxyhydantoin (NHH) carbamate that selectively inhibits the Wnt-deacylating enzyme NOTUM (IC50 13 nM) and preserves Wnt3A signaling in the presence of NOTUM.

  • Selective NOTUM inhibitor with IC50 of 13 nM.
  • Preserves Wnt3A signaling in the presence of NOTUM.
  • Molecular formula C22H21ClN4O5; molecular weight 456.88.
  • Purity 98.5% with supporting analytical documentation (COA, H NMR, LCMS, SDS).
  • Available as a 1 mg solid and as a 10 mM solution in DMSO.

Catalog No. 50-002-31459 Supplier Medchemexpress LLC Supplier No. HY1228321MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ABC99 is an N-hydroxyhydantoin (NHH) carbamate that selectively inhibits the Wnt-deacylating enzyme NOTUM (IC50 13 nM) and preserves Wnt3A signaling in the presence of NOTUM.

  • Selective NOTUM inhibitor with IC50 of 13 nM.
  • Preserves Wnt3A signaling in the presence of NOTUM.
  • Molecular formula C22H21ClN4O5; molecular weight 456.88.
  • Purity 98.5% with supporting analytical documentation (COA, H NMR, LCMS, SDS).
  • Available as a 1 mg solid and as a 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.