Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology ABCA1 Monoclonal Antibody, Rabbit Host, Human/Mouse/Rat Reactivity, 100 μL
SDP

Catalog No. 502729757
Encompass

This antibody recognizes endogenous levels of total ABCA1 protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Ser1262 of the human ABCA1 protein. The protein functions as a membrane-residing transporter that regulates phospholipid and cholesterol homeostasis in an ATP-dependent manner. This product is intended for research use only.

  • Host species: rabbit
  • Reactivity: human, mouse, and rat
  • Applications: Western blotting, immunoprecipitation
  • Molecular weight: 254 kDa
  • Storage: supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide; store at -20°C

Catalog No. 50-272-9757 Supplier Cell Signaling Technology Supplier No. 96292S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This antibody recognizes endogenous levels of total ABCA1 protein. It is produced by immunizing animals with a synthetic peptide corresponding to residues surrounding Ser1262 of the human ABCA1 protein. The protein functions as a membrane-residing transporter that regulates phospholipid and cholesterol homeostasis in an ATP-dependent manner. This product is intended for research use only.

  • Host species: rabbit
  • Reactivity: human, mouse, and rat
  • Applications: Western blotting, immunoprecipitation
  • Molecular weight: 254 kDa
  • Storage: supplied in 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and less than 0.02% sodium azide; store at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.